Anti-SFTPD Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological

90003-R001-100 90003-R001-50
103661-772EA 214.31 USD
103661-772 103597-538
Anti-SFTPD Rabbit Monoclonal Antibody [clone: 001]
Antibodies
Anti-Surfactant protein D Anti

Type: Primary
Antigen: SFTPD
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR