Anti-p53 Rabbit Monoclonal Antibody [clone: 002]

Supplier: Sino Biological

90001-R002-50 90001-R002-100
103597-532EA 128.15 USD
103597-532 103661-758
Anti-p53 Rabbit Monoclonal Antibody [clone: 002]
Antibodies
Anti-p53 Antibody

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: Clone: 002
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR