Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA protein Rabbit Monoclonal Antibody [clone: 133]

Supplier: Sino Biological

86001-RM02-100 86001-RM02-50
103662-404EA 312.31 USD InStock
103662-404 103597-526
Anti-HA protein Rabbit Monoclonal Antibody [clone: 133]
Antibodies
Influenza A Virus Hemagglutini

Type: Primary
Antigen: HA protein
Clonality: Monoclonal
Clone: Clone: 133
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: H1N1 (A/California/04/2009) ;H1N1 (A/California/07/2009) ;

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty