Anti-EphB3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
81593-T36-200 81593-T36-100 81593-T36-50
103637-804EA 142.96 USD
103637-804 103597-522 103597-520
Anti-EphB3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat EphB3 / HEK2 / Eph Receptor B3 (rR EphB3 / HEK2 / Eph Receptor B3; Catalog#81593-R08H; NP_001099338.1; Met1-Thr537). EphB3 / HEK2 / Eph Receptor B3 specific IgG was purified by Rat EphB3 / HEK2 / Eph Receptor B3 affinity chromatography.

Type: Primary
Antigen: EphB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR