Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bikunin Rabbit Polyclonal Antibody

Supplier: Sino Biological

81299-T60-100 81299-T60-200 81299-T60-50
103597-510EA 160.46 USD InStock
103597-510 103597-512 103637-800
Anti-Bikunin Rabbit Polyclonal Antibody
Antibodies
Anti-Bikunin Antibody

Type: Primary
Antigen: Bikunin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty