Anti-AMBP Rabbit Polyclonal Antibody

Supplier: Sino Biological
81299-RP01-200 81299-RP01-400 81299-RP01-100
103661-746EA 562.97 USD
103661-746 103661-742 103661-744
Anti-AMBP Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Protein HC / Bikunin / Ambp (rR Protein HC / Bikunin / Ambp; Catalog#81299-R08H; NP_037033.1; Met1-Ala202). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Bikunin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR