Anti-CTLA4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
81069-T16-200 81069-T16-50 81069-T16-100
103637-790EA 142.96 USD
103637-790 103597-492 103597-490
Anti-CTLA4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CTLA-4/CD152 (rR CTLA-4/CD152; Catalog#81069-R08H; NP_113862.1; Met1-Asp161). CTLA-4/CD152 specific IgG was purified by Rat CTLA-4/CD152 affinity chromatography.

Type: Primary
Antigen: CTLA-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR