Anti-CTLA4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
81069-RP01-100 81069-RP01-400 81069-RP01-200
103661-734EA 562.97 USD
103661-734 103661-730 103661-732
Anti-CTLA4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CTLA-4 / CD152 (Catalog#81069-R08H; NP_113862.1; Met1-Asp161). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CTLA-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR