Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STX7 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80948-T16-50 80948-T16-200 80948-T16-100
103637-786EA 106.62 USD InStock
103637-786 103597-484 103597-482
Anti-STX7 Rabbit Polyclonal Antibody
Antibodies
Anti-Syntaxin 7 Antibody

Type: Primary
Antigen: STX7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty