Anti-CPA2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80715-T48-100 80715-T48-200 80715-T48-50
103597-474EA 352.03 USD
103597-474 103637-782 103597-472
Anti-CPA2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Carboxypeptidase A2 / CPA2 (rR Carboxypeptidase A2 / CPA2; Catalog#80715-R08H; NP_001013101.2; Met1-Tyr417). Carboxypeptidase A2 / CPA2 specific IgG was purified by Rat Carboxypeptidase A2 / CPA2 affinity chromatography.

Type: Primary
Antigen: CPA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR