Anti-NPC2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80706-T56-200 80706-T56-100 80706-T56-50
103637-778EA 142.96 USD
103637-778 103597-466 103597-464
Anti-NPC2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat NPC2 (rR NPC2; Catalog#80706-R08H; CAD56199.1; Met1-Gly152). NPC2 specific IgG was purified by Rat NPC2 affinity chromatography.

Type: Primary
Antigen: NPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR