Anti-NPC2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80706-RP01-400 80706-RP01-200 80706-RP01-100
103661-706EA 106.43 USD
103661-706 103661-708 103661-710
Anti-NPC2 Rabbit Polyclonal Antibody
Antibodies
Anti-NPC2 Antibody

Type: Primary
Antigen: NPC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR