Anti-LDHA Rabbit Polyclonal Antibody

Supplier: Sino Biological

80664-RP01-400 80664-RP01-200 80664-RP01-100
103661-704EA 214.31 USD
103661-704 103661-702 103661-700
Anti-LDHA Rabbit Polyclonal Antibody
Antibodies
Anti-LDHA Antibody

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR