Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COPZ1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80636-T52-200 80636-T52-100 80636-T52-50
103597-454EA 214.31 USD InStock
103597-454 103597-452 103637-772
Anti-COPZ1 Rabbit Polyclonal Antibody
Antibodies
Anti-COPZ1 Antibody

Type: Primary
Antigen: COPZ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty