Anti-COPZ1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80636-RP01-100 80636-RP01-200 80636-RP01-400
103661-694EA 106.62 USD
103661-694 103661-696 103661-698
Anti-COPZ1 Rabbit Polyclonal Antibody
Antibodies
Anti-COPZ1 Antibody

Type: Primary
Antigen: COPZ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR