Anti-HAAO Rabbit Polyclonal Antibody

Supplier: Sino Biological

80627-T16-200 80627-T16-50 80627-T16-100
103597-446EA 214.31 USD
103597-446 103637-768 103597-444
Anti-HAAO Rabbit Polyclonal Antibody
Antibodies
Anti-HAAO Antibody

Type: Primary
Antigen: HAAO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR