Anti-Transferrin Rabbit Polyclonal Antibody

Supplier: Sino Biological

80586-RP01-200 80586-RP01-400 80586-RP01-100
103662-380EA 160.46 USD
103662-380 103662-382 103662-378
Anti-Transferrin Rabbit Polyclonal Antibody
Antibodies
Anti-Transferrin Antibody

Type: Primary
Antigen: Transferrin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR