Anti-AK1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80583-RP01-200 80583-RP01-100 80583-RP01-400
103661-680EA 351.49 USD
103661-680 103662-370 103661-678
Anti-AK1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat AK1/Adenylate kinase 1 (rR AK1/Adenylate kinase 1; Catalog#80583-R07E; NP_077325.2; Met1-Lys194). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: AK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR