Anti-ENO1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80578-T52-200 80578-T52-50 80578-T52-100
103597-424EA 352.03 USD
103597-424 103637-760 103597-422
Anti-ENO1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat ENO1 / Enolase 1 / alpha-enolase (Catalog#80578-R07E; NP_036686.2; Met1-Lys434). ENO1 / Enolase 1 / alpha-enolase specific IgG was purified by Rat ENO1 / Enolase 1 / alpha-enolase affinity chromatography.

Type: Primary
Antigen: ENO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR