Anti-ECM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80574-T16-200 80574-T16-100 80574-T16-50
103597-420EA 214.31 USD
103597-420 103597-418 103637-758
Anti-ECM1 Rabbit Polyclonal Antibody
Antibodies
Anti-ECM1 Antibody

Type: Primary
Antigen: ECM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR