Anti-SerpinA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80476-T16-200 80476-T16-50 80476-T16-100
103596-920EA 352.03 USD
103596-920 103637-754 103596-918
Anti-SerpinA1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat SerpinA1 (rR SerpinA1; Catalog#80476-R08H; NP_071964.2; Met1-Arg411). SerpinA1 specific IgG was purified by Rat SerpinA1 affinity chromatography.

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR