Anti-IL17F Rabbit Polyclonal Antibody

Supplier: Sino Biological
80462-T48-100 80462-T48-50 80462-T48-200
103597-414EA 352.03 USD
103597-414 103637-750 103597-412
Anti-IL17F Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL-17F (rR IL-17F; Catalog#80462-R08B; NP_001015011.1; Met1-Ala153). IL-17F specific IgG was purified by Rat IL-17F affinity chromatography.

Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR