Anti-EGF Mouse Monoclonal Antibody [clone: 02]

Supplier: Sino Biological
80446-MM02-200 80446-MM02-100 80446-MM02-50
103662-346EA 352.03 USD
103662-346 103662-344 103597-388
Anti-EGF Mouse Monoclonal Antibody [clone: 02]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat Epidermal Growth Factor / EGF (rR Epidermal Growth Factor / EGF; Catalog#80446-RNAE; NP_036974.1; Asn974-Arg1026). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EGF
Clonality: Monoclonal
Clone: 02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR