Anti-PAI-1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80442-RP01-400 80442-RP01-100 80442-RP01-200
103662-342EA 214.31 USD
103662-342 103662-340 103662-338
Anti-PAI-1 Rabbit Polyclonal Antibody
Antibodies
Anti-PAI-1 Antibody

Type: Primary
Antigen: PAI-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR