Anti-DDR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80440-T40-50 80440-T40-200 80440-T40-100
103637-738EA 99 USD
103637-738 103597-380 103597-378
Anti-DDR1 Rabbit Polyclonal Antibody
Antibodies
Anti-DDR1 Antibody

Type: Primary
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR