Anti-PDGFRA Rabbit Polyclonal Antibody

Supplier: Sino Biological
80433-RP01-400 80433-RP01-100 80433-RP01-200
103661-646EA 562.97 USD
103661-646 103661-642 103661-644
Anti-PDGFRA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat PDGFRA / CD140a (rR PDGFRA / CD140a; Catalog#80433-R08H; NP_036934.1; Met1-Glu523). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PDGFRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR