Anti-LAYN Rabbit Polyclonal Antibody

Supplier: Sino Biological

80431-T24-100 80431-T24-50 80431-T24-200
103597-370EA 160.46 USD
103597-370 103637-734 103597-372
Anti-LAYN Rabbit Polyclonal Antibody
Antibodies
Anti-LAYN Antibody

Type: Primary
Antigen: LAYN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR