Anti-LAYN Rabbit Polyclonal Antibody

Supplier: Sino Biological
80431-RP01-100 80431-RP01-400 80431-RP01-200
103661-636EA 351.49 USD
103661-636 103661-638 103661-640
Anti-LAYN Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Layilin / LAYN (Catalog#80431-R08H; NP_001178926.1; Met1-Glu224). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LAYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR