Anti-MOG Rabbit Polyclonal Antibody

Supplier: Sino Biological

80425-T56-100 80425-T56-200 80425-T56-50
103597-360EA 160.46 USD
103597-360 103597-362 103637-730
Anti-MOG Rabbit Polyclonal Antibody
Antibodies
Anti-MOG Antibody

Type: Primary
Antigen: MOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR