Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B2M Rabbit Monoclonal Antibody [clone: 035]

Supplier: Sino Biological

80423-R035-1 80423-R035-100 80423-R035-50
103637-726EA 1281.54 USD InStock
103637-726 103661-628 103597-354
Anti-B2M Rabbit Monoclonal Antibody [clone: 035]
Antibodies
Anti-Beta-2 microglobulin Anti

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: Clone: 035
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty