Anti-CLM-9 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80413-RP01-100 80413-RP01-400 80413-RP01-200
103661-622EA 106.62 USD
103661-622 103661-624 103661-620
Anti-CLM-9 Rabbit Polyclonal Antibody
Antibodies
Anti-CLM-9 Antibody

Type: Primary
Antigen: CLM-9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR