Anti-CD142 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80407-T16-200 80407-T16-50 80407-T16-100
103597-346EA 214.31 USD
103597-346 103637-720 103597-344
Anti-CD142 Rabbit Polyclonal Antibody
Antibodies
Anti-Tissue Factor Antibody

Type: Primary
Antigen: CD142
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR