Anti-F3 Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological
80407-MM01-100 80407-MM01-200 80407-MM01-50 80407-MM01-1
103637-718EA 1432.9 USD
103637-718 103597-342 103662-324 103662-322
Anti-F3 Mouse Monoclonal Antibody [clone: 01]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat Coagulation Factor III / Tissue Factor / CD142 (rR Coagulation Factor III / Tissue Factor / CD142; Catalog#80407-R08H; Q66HI4; Met1-Glu252). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD142
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR