Anti-NTRK1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80404-T24-200 80404-T24-50 80404-T24-100
103597-338EA 270.87 USD
103597-338 103637-716 103597-340
Anti-NTRK1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat TrkA / NTRK1 (Catalog#80404-R08H; NP_067600.1; Met1-Pro418). TrkA / NTRK1 specific IgG was purified by Rat TrkA / NTRK1 affinity chromatography.

Type: Primary
Antigen: NTRK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR