Anti-ART4 Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological
80383-MM01-100 80383-MM01-200 80383-MM01-50
103661-606EA 270.87 USD
103661-606 103661-608 103597-322
Anti-ART4 Mouse Monoclonal Antibody [clone: 01]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD297 (Catalog#80383-R08H; NP_001166980.1; Met1-Lys269). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.

Type: Primary
Antigen: ART4
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR