Anti-B7-H3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80380-T52-100 80380-T52-50 80380-T52-200
103597-318EA 160.46 USD
103597-318 103637-708 103597-320
Anti-B7-H3 Rabbit Polyclonal Antibody
Antibodies
Anti-B7-H3 Antibody

Type: Primary
Antigen: B7-H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR