Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 06]

Supplier: Sino Biological

80374-MM06-P-25 80374-MM06-P-100
103630-266EA 16.15 USD InStock
103630-266 103632-454
Anti-CD5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 06]
Antibodies
Anti-CD5 Antibody (PE)

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: Clone: 06
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty