Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 06]

Supplier: Sino Biological

80374-MM06-A-25 80374-MM06-A-100
103630-262EA 53.85 USD
103630-262 103632-450
Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 06]
Antibodies
Anti-CD5 Antibody (APC)

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: Clone: 06
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR