Anti-CD5 Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological
80374-MM06-100 80374-MM06-50 80374-MM06-200
103662-302EA 352.03 USD
103662-302 103597-310 103662-300
Anti-CD5 Mouse Monoclonal Antibody [clone: 06]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD5/Cluster of Differentiation 5 (rR CD5/Cluster of Differentiation 5; Catalog#80374-R08H; P51882; Met1-Pro368). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR