Anti-CD226 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80369-RP01-100 80369-RP01-200 80369-RP01-400
103661-590EA 270.2 USD
103661-590 103661-592 103661-594
Anti-CD226 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD226 / DNAM-1 (rR CD226 / DNAM-1; Catalog#80369-R08H; NP_001100840.1; Met1-Ile265). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD226
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR