Anti-GGT1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80364-T24-50 80364-T24-100 80364-T24-200
103637-702EA 106.62 USD
103637-702 103597-298 103597-300
Anti-GGT1 Rabbit Polyclonal Antibody
Antibodies
Anti-GGT1 Antibody

Type: Primary
Antigen: GGT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR