Anti-PROCR Rabbit Polyclonal Antibody

Supplier: Sino Biological
80362-RP01-100 80362-RP01-200 80362-RP01-400
103661-580EA 270.2 USD
103661-580 103661-582 103661-578
Anti-PROCR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat Epcr / PROCR (Catalog#80362-R08H; Met1-Ser213). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EPCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR