Anti-IL12B Rabbit Polyclonal Antibody

Supplier: Sino Biological
80352-T24-100 80352-T24-200 80352-T24-50
103597-286EA 352.03 USD
103597-286 103597-284 103637-696
Anti-IL12B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat IL12B/IL-12B (Catalog#80352-R08H; G3V9Y5; Met1-Ser335). IL12B/IL-12B specific IgG was purified by Rat IL12B/IL-12B affinity chromatography.

Type: Primary
Antigen: IL12B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR