Anti-BST1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80351-RP02-50 80351-RP02-200 80351-RP02-100
103597-272EA 142.96 USD
103597-272 103661-566 103661-568
Anti-BST1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat BST-1/BST1 (rR BST-1/BST1; Catalog#80351-R08H; Q63072; Met1-Glu293). BST-1/BST1 specific IgG was purified by Rat BST-1/BST1 affinity chromatography.

Type: Primary
Antigen: CD157
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR