Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-THBD Rabbit Polyclonal Antibody

Supplier: Sino Biological

80348-T24-50 80348-T24-200 80348-T24-100
103637-688EA 106.62 USD InStock
103637-688 103597-262 103597-260
Anti-THBD Rabbit Polyclonal Antibody
Antibodies
Anti-Thrombomodulin Antibody

Type: Primary
Antigen: THBD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty