Anti-LILRA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80334-RP01-200 80334-RP01-400 80334-RP01-100
103661-558EA 562.97 USD
103661-558 103661-554 103661-556
Anti-LILRA5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat LILRA5 (rR LILRA5; Catalog#80334-R08H; D4A6Y0; Met1-Asn248). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LILRA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR