Anti-LILRA5 Mouse Monoclonal Antibody [clone: 02]

Supplier: Sino Biological
80334-MM02-50 80334-MM02-200 80334-MM02-1 80334-MM02-100
103637-680EA 1432.9 USD
103637-680 103597-242 103661-550 103661-552
Anti-LILRA5 Mouse Monoclonal Antibody [clone: 02]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat LILRA5 (rR LILRA5; Catalog#80334-R08H; D4A6Y0; Met1-Asn248). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LILRA5
Clonality: Monoclonal
Clone: 02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR