Anti-CD83 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80333-T48-100 80333-T48-50 80333-T48-200
103597-238EA 270.87 USD
103597-238 103637-678 103597-240
Anti-CD83 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat CD83 (Catalog#80333-R08H; B2GV95; Met1-Ala134). CD83 specific IgG was purified by Rat CD83 affinity chromatography.

Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR