Anti-CD83 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80333-RP01-200 80333-RP01-400 80333-RP01-100
103661-544EA 160.46 USD
103661-544 103661-548 103661-546
Anti-CD83 Rabbit Polyclonal Antibody
Antibodies
Anti-CD83 Antibody

Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR