Anti-CD83 Rabbit Monoclonal Antibody [clone: 004]

Supplier: Sino Biological
80333-R004-100 80333-R004-50
103597-236EA 150.18 USD
103597-236 103661-542
Anti-CD83 Rabbit Monoclonal Antibody [clone: 004]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD83 (rR CD83; Catalog#80333-R08H; B2GV95; Met1-Ala134).

Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: 004
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR